siRNA Details
Gene Symbol: THUMPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304550.1 | NP_001291479.1 | 2 | 554 | ||
NM_017736.4 | NP_060206.2 | 2 | 554 | |||
XM_011545891.1 | XP_011544193.1 | 2 | 156 | |||
XM_017023433.1 | XP_016878922.1 | 2 | 496 | |||
GenBank mRNA | AK000281.1 | - | 2 | 408 | ||
BC000448.2 | - | 2 | 386 | 2821354 | ||
BX640898.1 | - | 2 | 674 |
