siRNA Details
Gene Symbol: BNC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317939.1 | NP_001304868.1 | 7 | 3606 | ||
NM_001317940.1 | NP_001304869.1 | 6 | 3456 | |||
NM_017637.5 | NP_060107.3 | 7 | 3676 | |||
XM_011517924.1 | XP_011516226.1 | 8 | 3741 | |||
XM_017014816.1 | XP_016870305.1 | 11 | 6127 | |||
XM_017014817.1 | XP_016870306.1 | 9 | 4201 | |||
XM_017014818.1 | XP_016870307.1 | 9 | 4191 | |||
XM_017014819.1 | XP_016870308.1 | 8 | 4134 | |||
XM_017014820.1 | XP_016870309.1 | 8 | 4128 | |||
XM_017014821.1 | XP_016870310.1 | 7 | 4048 | |||
XM_017014822.1 | XP_016870311.1 | 7 | 3737 | |||
XM_017014823.1 | XP_016870312.1 | 7 | 8506 | |||
XM_017014824.1 | XP_016870313.1 | 6 | 4058 | |||
XM_017014825.1 | XP_016870314.1 | 10 | 4233 | |||
XM_017014829.1 | XP_016870318.1 | 7 | 4138 | |||
GenBank mRNA | AK000050.1 | - | 2 | 1472 | ||
AK025882.1 | - | 2 | 1205 | |||
AK092247.1 | - | 4 | 3148 | |||
AK131398.1 | - | 7 | 3566 | |||
AY438376.1 | - | 5 | 3321 | |||
BX641030.1 | - | 2 | 2706 | |||
CR933649.1 | - | 7 | 3563 |
