siRNA Details
Gene Symbol: Olr1260 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000962.1 | NP_001000962.1 | 1 | 777 |
Gene Symbol: LOC100912388 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749618.1 | XP_003749666.1 | 1 | 777 |
Gene Symbol: LOC100912432 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749619.1 | XP_003749667.1 | 777 |
