siRNA Details
Gene Symbol: SNX29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522743.2 | XP_011521045.1 | 22 | 3376 | ||
XM_011522744.2 | XP_011521046.1 | 21 | 3136 | |||
GenBank mRNA | BC029857.1 | - | 9 | 1499 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
