siRNA Details
Gene Symbol: CNNM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017649.4 | NP_060119.3 | 8 | 3478 | ||
NM_199076.2 | NP_951058.1 | 7 | 3412 | |||
GenBank mRNA | AF216962.1 | - | 8 | 2690 | ||
AK000071.1 | - | 7 | 1881 | |||
AK023066.1 | - | 8 | 2592 | |||
BX113753.1 | - | 1 | 41 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
