siRNA Details
Gene Symbol: Fus | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012137.1 | NP_001012137.1 | 2, 3 | 97 | ||
XM_006230290.2 | XP_006230352.1 | 2, 3 | 124 | |||
GenBank mRNA | BC087153.1 | - | 2, 3 | 97 | 7132489 |
