siRNA Details
Gene Symbol: TCEAL5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012979.2 | NP_001012997.1 | 3 | 915 | ||
GenBank mRNA | AF086094.1 | - | 1 | 17 | ||
BC132922.1 | - | 3 | 792 | 40146944 | ||
BC132924.1 | - | 3 | 792 | 40146946 |
