siRNA Details
Gene Symbol: Mospd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014107.1 | NP_001014129.1 | 4, 5 | 397 | ||
XM_006257631.2 | XP_006257693.1 | 3, 4 | 443 | |||
XM_006257632.2 | XP_006257694.1 | 4, 5 | 482 | |||
XM_006257633.2 | XP_006257695.1 | 3, 4 | 440 | |||
XM_006257634.2 | XP_006257696.1 | 3, 4 | 440 | |||
GenBank mRNA | BC086521.1 | - | 4, 5 | 397 | 7302680 |
Gene Symbol: LOC686087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006251400.2 | XP_006251462.1 | 1 | 415 | ||
XM_008770405.1 | XP_008768627.1 | 1 | 415 |
