siRNA Details
Gene Symbol: Magix | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014109.1 | NP_001014131.1 | 6 | 1398 | ||
GenBank mRNA | BC078960.1 | - | 6 | 1398 | 7110859 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Magix | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014109.1 | NP_001014131.1 | 6 | 1398 | ||
GenBank mRNA | BC078960.1 | - | 6 | 1398 | 7110859 |