siRNA Details
Gene Symbol: VPS50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257998.1 | NP_001244927.1 | 14, 15 | 1242 | ||
NM_017667.3 | NP_060137.2 | 13, 14 | 1214 | |||
XM_011516395.2 | XP_011514697.1 | 13, 14 | 1204 | |||
XM_011516396.2 | XP_011514698.1 | 13, 14 | 1211 | |||
GenBank mRNA | AB058764.2 | - | 13, 14 | 1160 | ||
AB100163.1 | - | 13, 14 | 1171 | |||
AK000104.1 | - | 5, 6 | 446 | |||
AK055965.1 | - | 14, 15 | 1178 | |||
AK126478.1 | - | 14, 15 | 1216 | |||
AK299582.1 | - | 3, 4 | 335 | |||
AL832393.1 | - | 6, 7 | 794 | |||
AL833112.1 | - | 12, 13 | 1082 | |||
BC152424.1 | - | 13, 14 | 1160 | 8862538 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
