siRNA Details
Gene Symbol: Gin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025012.1 | NP_001020183.1 | 7 | 1249 | ||
XM_006245604.1 | XP_006245666.1 | 6 | 1172 | |||
XM_006245605.2 | XP_006245667.1 | 7 | 1668 | |||
XM_008767377.1 | XP_008765599.1 | 7 | 4844 | |||
XM_008767378.1 | XP_008765600.1 | 7 | 4504 | |||
GenBank mRNA | BC082058.1 | - | 7 | 1249 | 7132420 |
