siRNA Details
Gene Symbol: RGD1305807 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025276.1 | NP_001020447.1 | 4 | 270 | ||
XM_006238038.2 | XP_006238100.1 | 4 | 512 | |||
XM_006238039.1 | XP_006238101.1 | 3 | 187 | |||
GenBank mRNA | BC097368.1 | - | 4 | 270 | 7443399 |
