siRNA Details
Gene Symbol: RGD1305807 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025276.1 | NP_001020447.1 | 16 | 1465 | ||
XM_006238038.2 | XP_006238100.1 | 15 | 1578 | |||
XM_006238039.1 | XP_006238101.1 | 15 | 1382 | |||
GenBank mRNA | BC097368.1 | - | 16 | 1465 | 7443399 |

More Information
Other Species Targeted

Gene Ontology Categories:
