siRNA Details
Gene Symbol: ZNF518B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053042.2 | NP_444270.2 | 3 | 1322 | ||
XM_005248193.2 | XP_005248250.1 | 2 | 1157 | |||
XM_017008784.1 | XP_016864273.1 | 2 | 3780 | |||
XM_017008785.1 | XP_016864274.1 | 3 | 1548 | |||
XM_017008786.1 | XP_016864275.1 | 2 | 1382 | |||
GenBank mRNA | AB051516.1 | - | 1 | 674 | ||
AL833306.1 | - | 1 | 858 |
