siRNA Details
Gene Symbol: DEF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242821.1 | NP_001229750.1 | 5 | 644 | ||
NM_001242822.1 | NP_001229751.1 | 6 | 738 | |||
NM_017702.3 | NP_060172.1 | 6 | 741 | |||
GenBank mRNA | AK000193.1 | - | 6 | 677 | ||
BC015482.2 | - | 5 | 664 | |||
BC105592.1 | - | 5 | 584 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
