siRNA Details
Gene Symbol: WDR55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005268469.2 | XP_005268526.1 | 8 | 3958 | ||
XM_017009600.1 | XP_016865089.1 | 8 | 5323 | |||
GenBank mRNA | BC002482.2 | - | 8 | 2762 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
