siRNA Details
Gene Symbol: Olr82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001282.1 | NP_001001282.1 | 1 | 847 | ||
XM_006229914.1 | XP_006229976.1 | 2 | 880 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Olr82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001282.1 | NP_001001282.1 | 1 | 847 | ||
XM_006229914.1 | XP_006229976.1 | 2 | 880 |