siRNA Details
Gene Symbol: TMEM104 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321264.1 | NP_001308193.1 | 10 | 981 | ||
NM_017728.3 | NP_060198.3 | 10 | 3941 | |||
XM_006721963.1 | XP_006722026.1 | 10 | 3912 | |||
XM_017024797.1 | XP_016880286.1 | 10 | 3880 | |||
GenBank mRNA | AK000262.1 | - | 10 | 964 | ||
AK074029.1 | - | 10 | 3901 | |||
AK296621.1 | - | 10 | 1004 | |||
AL137521.1 | - | 1 | 3053 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
