siRNA Details
Gene Symbol: OR4F15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001674.1 | NP_001001674.1 | 1 | 457 | ||
GenBank mRNA | BC137064.1 | - | 1 | 481 | 9021061 | |
BC137065.1 | - | 1 | 481 | 9021062 |
