siRNA Details
Gene Symbol: Krt83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003668.2 | NP_001003668.2 | 7 | 1217 | ||
GenBank mRNA | BC139321.1 | - | 7 | 1217 | 8862343 | |
BC139322.1 | - | 7 | 1217 | 8862344 |

More Information
Other Species Targeted

Gene Ontology Categories:
