siRNA Details
Gene Symbol: Gm5617 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004191.3 | NP_001004191.2 | 1 | 606 | ||
GenBank mRNA | AK007723.1 | - | 1 | 511 | ||
BC011344.1 | - | 1 | 501 | 3481870 | ||
CN838422.1 | - | 1 | 510 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
