siRNA Details
Gene Symbol: ZNF783 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195220.1 | NP_001182149.1 | 4 | 727 | ||
XM_011515701.2 | XP_011514003.1 | 4 | 655 | |||
GenBank mRNA | BG029497.1 | - | 3 | 298 | ||
BX361979.2 | - | 4 | 683 |
