siRNA Details
Gene Symbol: SPDYE3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004351.4 | NP_001004351.3 | 1 | 191 | ||
XM_011516234.2 | XP_011514536.1 | 1 | 1094 | |||
XM_011516235.2 | XP_011514537.1 | 1 | 1094 | |||
GenBank mRNA | BC056606.1 | - | 1 | 192 | 4838469 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
