siRNA Details
Gene Symbol: Pnet-ps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142086.1 | - | 1 | 1529 | ||
AY223547.1 | - | 1 | 234 | |||
BY299462.1 | - | 1 | 321 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
