siRNA Details
Gene Symbol: Ddx19b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005895.2 | NP_001005895.2 | 6 | 582 | ||
XM_008772534.1 | XP_008770756.1 | 6 | 496 | |||
GenBank mRNA | AY751756.1 | - | 6 | 524 | ||
EV774940.1 | - | 6 | 582 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
