siRNA Details
Gene Symbol: FLVCR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017791.2 | NP_060261.2 | 1 | 957 | ||
GenBank mRNA | AF456126.1 | - | 1 | 581 | ||
AK000378.1 | - | 1 | 905 | |||
AY260572.1 | - | 1 | 937 | |||
AY260577.1 | - | 1 | 937 | |||
BC019087.2 | - | 1 | 920 | 4866427 |
