siRNA Details
Gene Symbol: Ccdc149 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256059.1 | NP_001242988.1 | 12 | 3034 | ||
GenBank mRNA | AA615092.1 | - | 1 | 93 | ||
AK049058.1 | - | 1 | 614 | |||
BB396098.2 | - | 1 | 384 | |||
BC025881.1 | - | 1 | 709 | 5027889 |
