siRNA Details
Gene Symbol: TMEM260 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017799.3 | NP_060269.3 | 8, 9 | 1072 | ||
XM_006720176.1 | XP_006720239.1 | 7, 8 | 861 | |||
XM_011536851.1 | XP_011535153.1 | 8, 9 | 1074 | |||
XM_011536852.1 | XP_011535154.1 | 8, 9 | 1064 | |||
XM_011536853.1 | XP_011535155.1 | 9, 10 | 1242 | |||
XM_017021379.1 | XP_016876868.1 | 8, 9 | 1074 | |||
XM_017021380.1 | XP_016876869.1 | 7, 8 | 861 | |||
GenBank mRNA | AK000399.1 | - | 5, 6 | 470 | ||
AK023885.1 | - | 1, 2 | 586 | |||
AK056291.1 | - | 8, 9 | 1072 | |||
AK293099.1 | - | 8, 9 | 1039 | |||
AK295989.1 | - | 7, 8 | 861 | |||
BC121163.1 | - | 8, 9 | 981 | 40121557 | ||
BC121164.1 | - | 8, 9 | 981 | |||
BX647387.1 | - | 8, 9 | 844 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
