siRNA Details
Gene Symbol: C20orf202 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009612.2 | NP_001009612.1 | 1 | 112 | ||
GenBank mRNA | AK092524.1 | - | 1 | 112 | ||
BC127730.1 | - | 1 | 42 | |||
BC127731.1 | - | 1 | 42 | |||
BC141955.1 | - | 1 | 42 | |||
BC141956.1 | - | 1 | 42 | |||
BC150532.1 | - | 1 | 42 | |||
CR746175.1 | - | 1 | 54 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
