siRNA Details
Gene Symbol: C20orf202 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009612.2 | NP_001009612.1 | 2 | 966 | ||
GenBank mRNA | AK092524.1 | - | 2 | 966 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
