siRNA Details
Gene Symbol: Ccdc159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750449.3 | XP_003750497.1 | 10 | 1095 | ||
XM_006242678.2 | XP_006242740.1 | 10 | 1095 | |||
XM_006242679.2 | XP_006242741.1 | 10 | 1092 | |||
XM_006242680.2 | XP_006242742.1 | 10 | 1065 | |||
XM_006242681.2 | XP_006242743.1 | 10 | 1062 | |||
XM_006242682.2 | XP_006242744.1 | 10 | 995 | |||
XM_006242683.2 | XP_006242745.1 | 9 | 965 | |||
XM_006242684.2 | XP_006242746.1 | 10 | 1149 | |||
XM_006242685.2 | XP_006242747.1 | 9 | 958 | |||
XM_006242686.2 | XP_006242748.1 | 8 | 734 | |||
GenBank mRNA | BC088314.1 | - | 7 | 635 | 7324729 |
