siRNA Details
Gene Symbol: Ccdc159 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006242678.2 | XP_006242740.1 | 12 | 1455 | ||
XM_006242679.2 | XP_006242741.1 | 12 | 1452 | |||
XM_006242680.2 | XP_006242742.1 | 12 | 1425 | |||
XM_006242681.2 | XP_006242743.1 | 12 | 1422 | |||
XM_006242682.2 | XP_006242744.1 | 12 | 1355 | |||
XM_006242683.2 | XP_006242745.1 | 11 | 1325 | |||
XM_006242684.2 | XP_006242746.1 | 12 | 1509 | |||
XM_006242685.2 | XP_006242747.1 | 11 | 1318 | |||
XM_006242686.2 | XP_006242748.1 | 10 | 1094 | |||
GenBank mRNA | BC088314.1 | - | 9 | 995 | 7324729 |
