siRNA Details
Gene Symbol: RGD1359508 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009707.1 | NP_001009707.1 | 6 | 491 | ||
XM_006232306.2 | XP_006232368.1 | 6 | 767 | |||
XM_006232307.2 | XP_006232369.1 | 5 | 462 | |||
XM_006232308.2 | XP_006232370.1 | 6 | 553 | |||
GenBank mRNA | BC088236.1 | - | 6 | 491 | 7325191 |
