siRNA Details
Gene Symbol: EXD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104599.1 | 3 | 1113 | |||
GenBank mRNA | AK000440.1 | - | 3 | 1013 | ||
BC039252.1 | - | 3 | 1013 | |||
BC065002.1 | - | 3 | 983 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
