siRNA Details
Gene Symbol: C10orf62 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009997.2 | NP_001009997.2 | 1 | 435 | ||
GenBank mRNA | BC037252.1 | - | 1 | 401 | 5163660 | |
BC043281.1 | - | 1 | 886 | |||
BI826342.1 | - | 1 | 434 |
