siRNA Details
Gene Symbol: C9orf153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276366.1 | NP_001263295.1 | 3 | 285 | ||
NM_001276367.1 | NP_001263296.1 | 4 | 364 | |||
NM_001276368.1 | NP_001263297.1 | 4 | 303 | |||
GenBank mRNA | BC108665.1 | - | 4 | 342 | 5164660 | |
BG772848.1 | - | 3 | 290 | |||
BI827110.1 | - | 4 | 304 |
