siRNA Details
Gene Symbol: SLC41A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008486.1 | NP_001008486.1 | 2, 3 | 507 | ||
XM_005247559.1 | XP_005247616.1 | 3, 4 | 493 | |||
XM_005247564.1 | XP_005247621.1 | 3, 4 | 493 | |||
XM_011512945.1 | XP_011511247.1 | 2, 3 | 642 | |||
XM_017006705.1 | XP_016862194.1 | 3, 4 | 482 | |||
XM_017006708.1 | XP_016862197.1 | 2, 3 | 340 | |||
XM_017006709.1 | XP_016862198.1 | 2, 3 | 395 | |||
GenBank mRNA | AK000480.1 | - | 2, 3 | 338 | ||
AK022780.1 | - | 2, 3 | 336 | |||
AK307107.1 | - | 3, 4 | 537 | |||
AL560923.3 | - | 2, 3 | 519 | |||
BM547018.1 | - | 500 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
