siRNA Details
Gene Symbol: Lims2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012163.1 | NP_001012163.1 | 3, 4 | 423 | ||
XM_006254596.2 | XP_006254658.1 | 3, 4 | 560 | |||
XM_006254597.2 | XP_006254659.1 | 3, 4 | 598 | |||
XM_008772043.1 | XP_008770265.1 | 4, 5 | 585 | |||
GenBank mRNA | BC087108.1 | - | 3, 4 | 423 | 7193274 |
