siRNA Details
Gene Symbol: Lims2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012163.1 | NP_001012163.1 | 7 | 893 | ||
XM_006254596.2 | XP_006254658.1 | 7 | 1030 | |||
XM_006254597.2 | XP_006254659.1 | 7 | 1068 | |||
XM_008772043.1 | XP_008770265.1 | 8 | 1055 | |||
GenBank mRNA | BC087108.1 | - | 7 | 893 | 7193274 | |
FQ226177.1 | - | 3 | 244 |
