siRNA Details
Gene Symbol: Rfx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012172.1 | NP_001012172.1 | 8 | 943 | ||
XM_006231233.2 | XP_006231295.1 | 9 | 1148 | |||
XM_006231235.2 | XP_006231297.1 | 8 | 1107 | |||
XM_008760340.1 | XP_008758562.1 | 7 | 916 | |||
XM_008760341.1 | XP_008758563.1 | 8 | 991 | |||
GenBank mRNA | BC085953.1 | - | 8 | 943 | 7134397 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001166414, NM_011265, BC017598, X76090, AK030008, and others.
Gene Ontology Categories:
