siRNA Details
Gene Symbol: Rfx3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012172.1 | NP_001012172.1 | 16 | 2252 | ||
XM_006231233.2 | XP_006231295.1 | 18 | 2612 | |||
XM_006231235.2 | XP_006231297.1 | 17 | 2571 | |||
XM_008760340.1 | XP_008758562.1 | 16 | 2380 | |||
GenBank mRNA | BC085953.1 | - | 16 | 2252 | 7134397 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_001166414, NM_011265, BC017598, AK132267, AK140928.
Gene Ontology Categories:
