siRNA Details
Gene Symbol: Mccc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012177.1 | NP_001012177.1 | 11, 12 | 1216 | ||
XM_008760678.1 | XP_008758900.1 | 10, 11 | 1288 | |||
GenBank mRNA | BC083581.1 | - | 11, 12 | 1216 | 7129446 |
