siRNA Details
Gene Symbol: Rbms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012184.1 | NP_001012184.1 | 11 | 1464 | ||
XM_006234234.2 | XP_006234296.1 | 12 | 1269 | |||
XM_006234235.2 | XP_006234297.1 | 12 | 1562 | |||
XM_006234236.2 | XP_006234298.1 | 12 | 1361 | |||
XM_006234237.2 | XP_006234299.1 | 12 | 2121 | |||
XM_006234238.2 | XP_006234300.1 | 11 | 1221 | |||
XM_006234239.2 | XP_006234301.1 | 13 | 1759 | |||
XM_006234241.2 | XP_006234303.1 | 11 | 1311 | |||
XM_008761860.1 | XP_008760082.1 | 12 | 1850 | |||
GenBank mRNA | BC087094.1 | - | 11 | 1464 | 7191334 |
