siRNA Details
Gene Symbol: FAM120C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198456.2 | NP_940858.2 | 2 | 824 | ||
GenBank mRNA | AK000513.1 | - | 2 | 768 | ||
AY121803.1 | - | 2 | 824 | |||
BC016138.1 | - | 2 | 824 | |||
BC081532.1 | - | 2 | 525 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
