siRNA Details
Gene Symbol: Vrk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012194.1 | NP_001012194.1 | 9 | 1113 | ||
XM_006240530.2 | XP_006240592.1 | 9 | 939 | |||
XM_006240531.2 | XP_006240593.1 | 9 | 969 | |||
XM_006240532.2 | XP_006240594.1 | 9 | 1361 | |||
XM_006240533.2 | XP_006240595.1 | 9 | 848 | |||
XM_008764822.1 | XP_008763044.1 | 9 | 1361 | |||
GenBank mRNA | BC079130.1 | - | 9 | 1113 | 7127355 |
