siRNA Details
Gene Symbol: Rangap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001244855.1 | NP_001231784.1 | 16 | 2133 | ||
XM_006242100.2 | XP_006242162.1 | 16 | 3573 | |||
XM_006242101.2 | XP_006242163.1 | 16 | 2422 | |||
GenBank mRNA | BC082056.1 | - | 16 | 2041 | 7132303 | |
CX570737.1 | - | 1 | 35 | |||
FQ217036.1 | - | 16 | 2117 |
