siRNA Details
Gene Symbol: Klhdc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012203.1 | NP_001012203.1 | 7 | 900 | ||
XM_006244542.2 | XP_006244604.1 | 6 | 750 | |||
GenBank mRNA | BC079035.1 | - | 7 | 900 | 7114311 |
