siRNA Details
Gene Symbol: C1orf146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012425.1 | NP_001012425.1 | 6 | 548 | ||
XM_011541447.2 | XP_011539749.1 | 7 | 743 | |||
GenBank mRNA | BC130499.1 | - | 6 | 542 | 40146530 | |
BC171742.1 | - | 6 | 568 | |||
BU567193.1 | - | 6 | 627 |
