siRNA Details
Gene Symbol: C1orf146 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012425.1 | NP_001012425.1 | 3 | 185 | ||
XM_011541447.2 | XP_011539749.1 | 4 | 380 | |||
GenBank mRNA | BC130499.1 | - | 3 | 179 | 40146530 | |
BQ435566.1 | - | 4 | 403 | |||
BQ435801.1 | - | 4 | 403 | |||
BU567193.1 | - | 3 | 264 | |||
BU852979.1 | - | 403 |
