siRNA Details
Gene Symbol: Rxfp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012475.1 | NP_001012493.1 | 15, 16 | 1318 | ||
GenBank mRNA | AY906861.1 | - | 15, 16 | 1318 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_080468, AK034592, AK047861, BC120741, AF346501, and others.
Gene Ontology Categories:
